Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ANA 12
GREENER_CHOICE

Catalog No. 478110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-811-0 10 mg
47-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-811-0 Supplier Tocris Bioscience™ Supplier No. 4781/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

TrkB receptor antagonist

TrkB receptor antagonist. Demonstrates a 2-site mode of action, preventing activation by BDNF non-competitively (IC50 values are 45.6 nM and 41.1 μM for the high and low affinity sites respectively). Displays no effect on TrkA and TrkC in an assay of neurite outgrowth. Demonstrates antidepressant properties. Also exhibits anti-seizure effects in an ischemia-induced neonatal seizure model by blocking down-regulation of KCC2.

Chemical Identifiers

CAS 219766-25-3
Molecular Formula C22H21N3O3S

Specifications

Quantity 10 mg
Formula Weight 407.49
Purity >99%
Molecular Weight (g/mol) 407.49
Chemical Name or Material ANA 12
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.