Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Interscience Anabac

Click to view available options
Scent:
Apple
Coconut
Eucalyptus
Floral
Lemon
Orchard Peach
Vanilla
Description
Autoclave deodorant available in the following scents:
- Eucalyptus
- Apple
- Lemon
- Orchard Peach
- Coconut, 100% natural perfume, Ecocert Certified
- Vanilla, 100% natural perfume, Ecocert Certified
- Floral, 100% natural perfume, Ecocert Certified
Specifications
Specifications
Product Type | Autoclave Capsule |
Color-coded | Yes |
Form | Capsules |
Scent | Orchard Peach |
Sterility | Non-sterile |
Appearance | Clear Amber |
Autoclavable | Autoclavable |
Color | Orange |
Disposable | Single-use |
Filtered | Non-filtered |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction