Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience Anabac
SDP

Catalog No. p-200109774
Click to view available options
Scent:
Apple
Coconut
Eucalyptus
Floral
Lemon
Orchard Peach
Vanilla

Autoclave deodorant. Available in a variety of scents.

Autoclave deodorant available in the following scents:

  • Eucalyptus
  • Apple
  • Lemon
  • Orchard Peach
  • Coconut, 100% natural perfume, Ecocert Certified
  • Vanilla, 100% natural perfume, Ecocert Certified
  • Floral, 100% natural perfume, Ecocert Certified

Specifications

Product Type Autoclave Capsule
Color-coded Yes
Form Capsules
Scent Eucalyptus
Sterility Non-sterile
Appearance Clear Amber
Autoclavable Autoclavable
Color Translucent Yellow
Disposable Single-use
Filtered Non-filtered
Flammable No
For Use With (Application) Autoclave Deodorant
Quantity 100/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.