Learn More
BINDER™ Analog Output
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers KBF 1020
Description
- For temperature and humidity values
- Output not adjustable
Specifications
Specifications
Description | Analog output 4-20 mA - for temperature and humidity values (output not adjustable) |
For Use With (Equipment) | BINDER Chambers KBF 1020 |
Material | Plastic |
Type | Analog Output |
Amperage | 4 to 20 mA |