Learn More
BINDER™ Analog Output
SDP
Catalog No. 08111118 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers KBF P 720
Catalog No. 08-111-118
Supplier: BINDER™ 80121738
Only null left
Add to Cart
Add to Cart
For Use With: KBF P 720
  • For temperature and humidity values
  • Output not adjustable
Specifications
Description Analog output 4-20 mA - for temperature and humidity values (output not adjustable)
For Use With (Equipment) BINDER Chambers KBF P 720
Material Plastic
Type Analog Output
Amperage 4 to 20 mA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.