Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Analog Output
Shop All BINDER Inc. Products

Click to view available options
Material:
Plastic
Description
- Analog output 4-20 mA for temperature values
- Output not adjustable
Specifications
Specifications
Description | Analog Output 4-20 mA - for Temperature Values |
For Use With (Equipment) | Units Series M |
Material | Plastic |
Weight (English) | 2.20 lb. |
Weight (Metric) | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction