Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANAPC2 Antibody (OTI1A6), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271860G
Description
ANAPC2 Monoclonal specifically detects ANAPC2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ANAPC2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANAPC2 | |
Full length recombinant protein of human ANAPC2 (NP_037498) produced in HEk293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A6 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
anaphase promoting complex subunit 2, anaphase-promoting complex subunit 2, APC2KIAA1406Cyclosome subunit 2, RP11-350O14.5 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
29882.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction