Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANAPC2 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503404

Description
APC2 is a cullin-related subunit of the anaphase-promoting complex (APC), or cyclosome. APC2 specifically interacts with APC11, another APC subunit.Specifications
ANAPC2 | |
Monoclonal | |
0.82 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UJX6 | |
ANAPC2 | |
Full length human recombit protein of human ANAPC2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1B12 | |
Unconjugated | |
ANAPC2 | |
APC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29882 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction