Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANAPC2 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503407S


Description
ANAPC2 Monoclonal antibody specifically detects ANAPC2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
ANAPC2 | |
Monoclonal | |
0.38 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UJX6 | |
Mouse | |
Affinity chromatography | |
RUO | |
29882 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI1H8 | |
Unconjugated | |
ANAPC2 | |
9230107K09Rik; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; Apc2; cyclosome subunit 2; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406 | |
Full length human recombinant protein of human ANAPC2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction