Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANAPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143952
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143952 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143952 Supplier Invitrogen™ Supplier No. PA5143952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human Jurkat whole cell, human HEPG2 whole cell, human HELA whole cell, monkey COS-7 whole cell, human A549 whole cell, human PC-3 whole cell, rat stomach tissue, rat testis tissue, rat lung tissue, rat PC-12 whole cell, mouse stomach tissue, mouse lung tissue, mouse RAW2647 whole cell. IHC: human rectal cancer tissue. ICC/IF: A431 cell. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANAPC2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ANAPC2
Gene Accession No. Q8BZQ7, Q9UJX6
Gene Alias 9230107K09Rik; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; Apc2; cyclosome subunit 2; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406
Gene Symbols ANAPC2
Host Species Rabbit
Immunogen E.coli-derived human ANAPC2 recombinant protein (Position: K51-R272).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29882, 99152
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.