Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Andes Virus Nucleocapsid Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572793
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572793 Supplier Invitrogen™ Supplier No. PA572793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Andes Virus Nucleocapsid Polyclonal Antibody for ELISA

Andes Virus Nucleocapsid is part of the virus internal structure that forms helical nucleocapsids of (-) ssRNA segments in small, medium and large viral segments. Andes virus is a type of hantavirus that is found in rodents in South America. People can become ill with Andes virus if they come in contact with infected rodents or their droppings while in South America, or less commonly if they have close contact with a person who is ill with Andes virus infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Andes Virus Nucleocapsid
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANDVsSgp1
Gene Alias ANDV
Gene Symbols ANDVsSgp1
Host Species Rabbit
Immunogen Andes virus nucleocapsid antibody for a 19 amino acid peptide near the amino terminus of the Andes virus nuclecapsid. The immunogen is within amino acids 130 - 180 of Andes Virus Nucleocapsid.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 991232
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.