Learn More
Eisco™ Andesite Specimen
SCI_ED
Catalog No. S24747 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24747 12/Pk.
S24746 1 Ea.
S26640 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24747 Supplier Eisco™ Supplier No. ESNG0040PK12
Add to Cart
Add to Cart
Andesite Specimen for introduction to igneous rock classification and identifying minerals within igneous rocks.
  • Abundant phenocrysts in fine-grained matrix offer great demonstration of igneous porphyry
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Texture and chemistry variation are useful for students learning about differing cooling rates
  • The fine-grained felsic matrix contains larger, identifiable hornblende crystals, indicating two different rates of cooling
  • Example of porphyry in igneous rocks
Specifications
Product Type Igneous Rock
For Use With (Application) Introduction to igneous rock classification and identifying minerals within igneous rocks
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Andesite