Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Androgen R/NR3C4 Antibody (2H8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280576
Description
Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Androgen R/NR3C4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA : 1:10000, Immunohistochemistry : 1:200-1:1000, Immunohistochemistry-Paraffin : 1:200-1:1000 | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2H8 | |
Unconjugated | |
PBS | |
AR | |
Purified recombinant fragment of human Androgen Receptor expressed in E. coli. [UniProt# P10275] | |
0.1 mL | |
Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
367 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction