Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Androgen R/NR3C4, p Ser210, p Ser213 Antibody (156C135.2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Androgen R/NR3C4 (p Ser210, p Ser213) |
---|---|
Clone | 156C135.2 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-4 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23109901
![]() |
Novus Biologicals
NBP2310990.1MG |
0.1 mg |
Each of 1 for $560.50
|
|
|||||
Description
Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Androgen R/NR3C4 (p Ser210, p Ser213) | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Primate | |
P10275 | |
367 | |
IgG1 κ | |
Protein G purified | |
In IGF-1 stimulated LNCaP cells (passage number 38), a ∼110 kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen. |
156C135.2 | |
Western Blot 1-4 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:20-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
AR | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title