Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Androgen R/NR3C4, p Ser94 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229527
Description
Androgen R/NR3C4 Polyclonal specifically detects Androgen R/NR3C4 in Human samples. It is validated for Western Blot.Specifications
Androgen R/NR3C4 (p Ser94) | |
Polyclonal | |
Western Blot 1:1000 | |
P10275 | |
AR | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser94 conjugated to KLH. Accession # P10275 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼110k AR protein phosphorylated at Ser94. Immunolabeling is blocked by preadsorption of antibody with the phospho-peptide that was used to generate the antibody but not by the corresponding dephospho-peptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Rabbit | |
110 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
367 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction