Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Androgen R/NR3C4 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC58762S100UG
This item is not returnable.
View return policy
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Flow CytometrySpecifications
Androgen R/NR3C4 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
367 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
2621E | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/106 cells | |
AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Synthetic Human Androgen R/NR3C4 peptide | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction