Learn More
Invitrogen™ Androgen Receptor Monoclonal Antibody (2H8)

Description
MA5-15598 targets Androgen receptor in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15598 immunogen is purified recombinant fragment of human AR expressed in E. Coli. MA5-15598 detects Androgen receptor which has a predicted molecular weight of approximately 110kDa.
Specifications
Specifications
| Antigen | Androgen Receptor |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2H8 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | AR |
| Gene Accession No. | P10275 |
| Gene Alias | AIS; Andr; androgen nuclear receptor variant 2; androgen receptor; androgen receptor (Testicular feminization), same as Tfm; androgen receptor splice variant 4b; Ar; AR8; AR-A; AW320017; DHTR; dihydrotestosterone receptor; HUMARA; HYSP1; KD; NR3C4; Nuclear receptor subfamily 3 group C member 4; RP11-383C12.1; SBMA; SMAX1; TFM; Variant AR45 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.