Learn More
Andwin Scientific Kettle Mantles for PYREX™ Reaction Kettles
SDP
Lower sections moldblown for breakage resistance
$456.55 - $629.10
Specifications
Product Type Mantle
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
11-847-15B
SDP
Encompass
View Documents
Andwin Scientific
TM 572
11-847B kettles
Each for $456.55
Only null left
Add to Cart
11-847-15E
SDP
Encompass
View Documents
Andwin Scientific
TM 580
11-847E Kettles
Each for $629.10
Only null left
Add to Cart
Description
Description
  • Heavy pressed covers have four openings to accommodate interchangeable standard taper joints, allowing quick assembly with condensers, stirrers and funnels
  • Glass kettles designed for organic reactions involving viscous materials
  • Wide mouth for easy cleaning of interior
  • Finely ground flanges for tight seal
  • 500 and 1000mL sizes have interchangeable covers with four standard taper 24/40 joints; 2000, 3000, 4000mL sizes have one standard taper 34/45 and three standard taper 24/40 joints
†Mantles available for other types of reaction kettles. Call 1-800-766-7000 for details.
Certifications
CSA certified
Specifications
Specifications
Mantle
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.