Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANGEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581567 Supplier Invitrogen™ Supplier No. PA581567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has specificity for Mouse ANGEL1.

ANGEL1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cis-Golgi network; cytosol; endoplasmic reticulum; nucleus; perinuclear region of cytoplasm; eukaryotic initiation factor 4E binding; protein domain specific binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGEL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ANGEL1
Gene Accession No. Q8VCU0
Gene Alias 1110030H02Rik; angel homolog 1; angel homolog 1 (Drosophila); angel1; angel1 {ECO:0000312; Ccr4e; EMBL:AAI66827.1}; KIAA0759; LOW QUALITY PROTEIN: protein angel homolog 1; mKIAA0759; protein angel homolog 1; RGD1306238
Gene Symbols ANGEL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Mouse ANGEL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68737
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.