Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Mouse anti-Mouse, Clone: C527, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1111
Description
Anti-Angiogenin, clone C527 is a mouse monoclonal antibody that detects Angiogenin and has been tested for use in ELISA, Immunofluorescence, and Western Blotting.
Specifications
Angiogenin | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Ang,Ang1,Rnase5,Rnase5a | |
Full length recombinant murine Angiogenin. | |
100 μg | |
Apoptosis and Cancer | |
NP_001155203 | |
Mouse | |
Purified |
ELISA, Immunofluorescence, Western Blot | |
C527 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone C527 detects Angiogenin in murine cells. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction