Learn More
Invitrogen™ Angiogenin Polyclonal Antibody

Description
Immunogen sequence: QDNSRYTHFL TQHYDAKPQG RDDRYCESIM RRRGLTSPCK DINTFIHGNK RSIKAICENK NGNPHRENLR ISKSSFQVTT CKLHGGSPWP PCQYRATAGF RNVVVACENG LPVHLDQSIF RRP; Positive Samples: LO2, HeLa, NCI-H460, B cells, Mouse liver, Rat liver; Cellular Location: Nucleus, basement membrane, extracellular matrix, extracellular space, nucleolus.
Specifications
Specifications
| Antigen | Angiogenin |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | ANG |
| Gene Accession No. | P03950, P21570 |
| Gene Alias | AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a |
| Gene Symbols | ANG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.