Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-1 Antibody (OTI8H2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270172JF646

Catalog No. N270172J646


Only null left
Add to Cart

Description

Description

Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Angiopoietin-1
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ANGPT1
Recombinant protein fragment corresponding to amino acids 198-498 of human Angiopoietin-1 (NP_001137) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI8H2
Western Blot, Immunohistochemistry
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003
Mouse
Affinity Purified
Angiogenesis, Cancer, Cell Cycle and Replication
284.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.