Learn More
Invitrogen™ Angiopoietin 1 Polyclonal Antibody

Description
Immunogen sequence: EKENLQGLVT RQTYIIQELE KQLNRATTNN SVLQKQQLEL MDTVHNLVNL CTKEGVLLKG GKREEEKPFR Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 90%.
Specifications
Specifications
| Antigen | Angiopoietin 1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Angpt1 |
| Gene Accession No. | Q15389 |
| Gene Alias | 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188 |
| Gene Symbols | Angpt1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.