Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Angiopoietin 1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523954

Catalog No. PIPA523954


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with bovine, mouse and porcine based on sequence homology.

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Angiopoietin 1
Polyclonal
Unconjugated
Angpt1
1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188
Rabbit
Antigen affinity chromatography, Protein A
RUO
11600, 284
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.34 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O08538, Q15389
Angpt1
KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human ANGPT1.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.