Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Angiopoietin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546968
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546968 Supplier Invitrogen™ Supplier No. PA546968
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) Angiopoietin-2, rhAngiopoietin-4, and rhAngiopoietin-like Factor/CDT6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiopoietin 1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Angpt1
Gene Accession No. O08538, Q15389
Gene Alias 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188
Gene Symbols Angpt1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 Ser20-Phe498.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11600, 284
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.