Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Angiopoietin 2/3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551083

Catalog No. PA551083


Only null left
Add to Cart

Description

Description

Angiopoietin 2 + 3 Polyclonal Antibody detects endogenous levels of Angiopoietin 2 + 3 protein.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Angiopoietin 2/3
Polyclonal
Unconjugated
ANGPTL2
AI593246; ANG3; ANG-5; angiopoietin 5; angiopoietin like 2; angiopoietin like 3; angiopoietin related protein 2; Angiopoietin-5; angiopoietin-like 2; angiopoietin-like 3; angiopoietin-like protein 2; Angiopoietin-like protein 3; angiopoietin-related protein 2; Angiopoietin-related protein 3; ANGPT3; ANGPT5; Angptl2; Angptl3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; ARP1; ARP2; AW260363; FHBL2; HARP; hypl; hypolipidemia; UNQ153/PRO179; UNQ170/PRO196
Rabbit
Antigen affinity chromatography
RUO
171100, 23452, 26360, 27329, 30924, 502970
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9R045, Q9R182, Q9UKU9, Q9Y5C1
ANGPTL2, ANGPTL3
Synthesized peptide derived from the Internal region of human Angptl1/2.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.