Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-2 Antibody (OTI3H7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270173UV
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Angiopoietin-2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANGPT2 | |
Recombinant protein fragment corresponding to amino acids 19-496 of human Angiopoietin-2 (NP_001138) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3H7 | |
Western Blot, Immunohistochemistry | |
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer | |
285.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction