Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin 2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990063
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0063 25 μL
50-194-3769 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0063 Supplier Rockland Immunochemicals Supplier No. 100401402S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Angiopoietin 2 Antibody antiserum is directed against mouse angiopoietin-2 and shows no reactivity with angiopoietin-1 from mouse-derived proteins. This reagent cross-reacts with human angiopoietin-2. Partially cross-reactivity is noted with human angiopoietin-1.

Angiopoietin-2 (ANGPT2) is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene.

Specifications

Antigen Angiopoietin 2
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 80 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2
Gene ANGPT2
Gene Accession No. O15123, O35608
Gene Alias Agpt2; ANG2; ANG-2; angiopoietin 2; Angiopoietin-2; angiopoietin-2a; angiopoietin-2B; ANGPT2; Tie2-ligand
Gene Symbols ANGPT2
Host Species Rabbit
Immunogen Anti-Angiopoietin 2 Antiserum was prepared by repeated immunizations with a synthetic peptide, corresponding to a region near the N-terminus of mouse angiopoietin-2 protein, conjugated to KLH using maleimide.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11601, 285
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.