Learn More
Invitrogen™ ANGPTL1 Monoclonal Antibody (OTI1B2)

Description
ANGPTL1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ANGPTL1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Angptl1 |
| Gene Accession No. | O95841 |
| Gene Alias | 2810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO188 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.