Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANGPTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552681 Supplier Invitrogen™ Supplier No. PA552681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKMATRYREL EVKYASLTDL VNNQSVMITL LEEQCLRIFS RQDTHVSPPL VQVVPQHIPN SQQYTPGLLG GNEIQRDPGY PRDLMPPPDL ATSPTKSPFK IPPVTFINEG PFKDCQQAKE AGHSVSGIYM IKPENSNGPM QLWCENS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Angptl1
Gene Accession No. O95841
Gene Alias 2810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO188
Gene Symbols Angptl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Angiopoietin 3. Recombinant protein control fragment (Product #RP-102169).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.