Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-4 Antibody (MM0090-11M11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212037AF594
Description
Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Angiopoietin-4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ANGPT4 | |
Human recombinant Angiopoietin-4 | |
0.1 mL | |
51378.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0090-11M11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction