Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Angiopoietin-4 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Angiopoietin-4 Polyclonal specifically detects Angiopoietin-4 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Angiopoietin-4 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Goat | |
Primate, Human | |
AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183 | |
ANGPT4 | |
IgG | |
Affinity Purified | |
Sequence Similarity: CPPGPEVSRDSNTLQRESLANPLHLGKLPTQQV 55-87. Human NP-057069 EASRDSNTLQRESLANPLRLEKLPTQQV 60-87 Macaque |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
PBS with 1 mg/ml BSA with 0.1% Sodium Azide | |
51378 | |
Synthetic peptide (CPPGPEVSRDSNTLQRESLANPLHLGKLPTQQV) corresponding to aa 55-87 of human Angiopoietin 4. | |
Primary | |
Store at -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title