Learn More
Angiopoietin-4 Antibody (RM0073-2H34), Janelia Fluor™ 646, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP121700JF646
Catalog No. NBP12700646
Only null left
Add to Cart
Description
Description
Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Angiopoietin-4
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ANGPT4
Mouse recombinant Angiopoietin-3
0.1 mL
51378
Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0073-2H34
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183
Rat
Protein A or G purified
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Angiopoietin-3.
Store at 4°C in the dark.
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.