Learn More
Novus Biologicals™ Angiopoietin-like Protein 2/ANGPTL2 Overexpression Lysate
Shop All R&D Systems Products

Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC21917 and based on accession number NM_1298. The protein contains a C-terminal DDK tag. |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens angiopoietin-like 2 (ANGPTL2), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cardiovascular Biology |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.