Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-like Protein 4/ANGPTL4 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$220.00 - $524.00
Specifications
Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Angiopoietin-like Protein 4/ANGPTL4 Polyclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse, Rat samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 4/ANGPTL4 | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Rat, Mouse | |
Q9Z1P8 | |
51129 | |
Synthetic peptide surrounding amino acid 84 of mouse FIAF (BAC25242). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
ANGPTL4 | |
IgG | |
Affinity Purified | |
It recognizes ∼50 kDa FIAF from samples of mouse and rat origins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title