Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-like protein 8/Betatrophin Antibody (OTI1B12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272046B
Description
Angiopoietin-like protein 8/Betatrophin Monoclonal specifically detects Angiopoietin-like protein 8/Betatrophin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Angiopoietin-like protein 8/Betatrophin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ANGPTL8 | |
| Recombinant protein fragment corresponding to amino acids 22-198 of human Angiopoietin-like protein 8/Betatrophin (NP_061157) produced in E.coli. | |
| 0.1 mL | |
| 55908.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1B12 | |
| Western Blot, Immunohistochemistry | |
| C19orf80, chromosome 19 open reading frame 80, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, PRO1185, PVPA599, TD26, UNQ599 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction