Learn More
3B Scientific™ Angiospermae Leaves Slide Set
Shop All 3B Scientific Products
Click to view available options
Quantity:
15
Description
- 1(d). Elodea, l.s. of stem tip showing apical meristem and origin of leaves
- 2(d). Leaves, monocot and dicot, Zea and Ranunculus, t.s.
- 3(c). Syringa, lilac, t.s. of typical dicot leaf
- 4(c). Iris, typical isobilateral leaf t.s.
- 5(c). Eucalyptus, a bifacial foliage leaf with schizogenous oil glands t.s.
- 6(d). Fagus, beech, t.s. of sun and shade leaves on one slide
- 7(c). Calluna, ling, t.s. of rolled leaf showing sunken stomata
- 8(c). Nerium oleander, t.s. of leaf showing sunken stomatal pits lined with protective hairs
- 9(c). Ficus elastica, rubber plant, t.s. of leaf showing cystoliths
- 10(c). Elodea, t.s. of leaf showing the simple structure of an aquatic leaf
- 11(c). Tulipa, tulip, epidermis w.m. showing stomata
- 12(d). Aesculus, t.s. of leaf bud with squama and embedded folded leaves
- 13(d). Drosera, sundew, w.m. of leaf with glandular hairs
- 14(d). Nepenthes, t.s. of pitcher with glands
- 15(d). Utricularia, bladderwort, w.m. of bladder
Specifications
Specifications
Type | Angiospermae Leaves |
Quantity | 15 |