Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiostatin Antibody (MM0094-6F34), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212042AF532
Description
Angiostatin Monoclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.Specifications
Angiostatin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein G purified | |
Biologically Active Proteins | |
5340.0 | |
Store at 4°C in the dark. |
Western Blot | |
MM0094-6F34 | |
Western Blot | |
PLG | |
Human recombinant Angiostatin | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction