Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin Converting Enzyme 1 (ACE) Monoclonal Antibody (OTI2C8), TrueMAB™, OriGene
SDP

Catalog No. 502543765
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3765 Supplier OriGene Supplier No. CF812211
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiotensin Converting Enzyme 1 (ACE) Monoclonal antibody specifically detects Angiotensin Converting Enzyme 1 (ACE) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Angiotensin Converting Enzyme 1 (ACE)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TERF1
Gene Accession No. P54274
Gene Alias ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA
Host Species Mouse
Immunogen Full length human recombinant protein of human TERF1 (NP_003209) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7013
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.