Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensin I Antibody (BGN/KA/4H), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062344AF594
Description
Angiotensin I Monoclonal specifically detects Angiotensin I in Human, Rat samples. It is validated for ELISA.Specifications
Angiotensin I | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
AGT | |
Synthetic peptide corresponding to amino acids 40-49 of the angiotensinogen precursor (Swiss-Prot entry P01019) conjugated to HMW carrier protein. | |
0.1 mL | |
183.0 | |
Human, Rat | |
IgG2a |
ELISA | |
BGN/KA/4H | |
ELISA | |
alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
Mouse | |
Protein A purified | |
Primary | |
NB100-62344 is specific for Angiotensin I and does not react with angiotensin II, III or angiotensinogen in ELISA. Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. Angiotensin I is important in the Renin-Angiotensin-Aldosterone System (RAAS). RAAS is responsible for the regulation of blood pressure and water retention in the body. Angiotensin I is produced from a reaction between the renal enzyme Renin and angiotensinogen. It is converted by Angiotensin Converting Enzyme (ACE) into a vasoactive form called Angiotensin II which is responsible for increasing blood pressure. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction