Learn More
Angiotensin I (human)
Description
Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Angiotensin I (human) stimulates the release of aldosterone, another hormone, from the adrenal cortex. It acts as a precursor to angiotensin II. It acts as endogenous peptide substrate for angiotensin converting enzyme (ACE), precursor to the vasoconstrictor peptide angiotensin II
Solubility
Soluble to 1 mg/ml in water.
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.

Specifications
Specifications
Color | White |
Quantity | 5 mg |
Solubility Information | Soluble to 1mg/ml in water. |
Formula Weight | 1296.5 |
Physical Form | Powder |
Chemical Name or Material | Angiotensin I (human) |
Safety and Handling
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.