Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensin II Antibody (KAA8), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320123AF647
Description
Angiotensin II Monoclonal antibody specifically detects Angiotensin II in Human, Rat samples. It is validated for Western BlotSpecifications
Angiotensin II | |
Monoclonal | |
Alexa Fluor 647 | |
AGT, alpha-1 antiproteinase, antitrypsin, angiotensin I, ANHU, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8 | |
Angiotensin II | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
KAA8 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
183 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction