Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiotensin II, MP Biomedicals™

Catalog No. p-7083079 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg

Angiotensin II Human From Synthetic, Powder

  • Angiotensin II plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.
  • Angiotensin II is a peptide hormone which plays an important role in the regulation of blood pressure.
  • It stimulates the release of aldosterone from the adrenal gland.It has strong vasoconstrictive effects.
TRUSTED_SUSTAINABILITY

Specifications

CAS 68521-88-0
Color Off-white
Content And Storage -20°C
Format Powder
Molecular Weight (g/mol) 1046
Quantity 5 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.