Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensin II/III Antibody (Ang II E7 (BGN/KA/4L)), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062346H
Description
Angiotensin II/III Monoclonal specifically detects Angiotensin II/III in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Angiotensin II/III | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AGT | |
Peptide sequence corresponding to angiotensin II, conjugated to a proprietary molecular weight carrier. Peptide sequence: DRVYIHPF | |
0.1 mL | |
Lipid and Metabolism | |
183 | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Ang II E7 (BGN/KA/4L) | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AGT angiotensinogen (serpin peptidase inhibitor, clade A, member 8), angiotensin I, angiotensin II, ANHU, SERPINA8 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This product reacts with Angiotensin II and III. Negligible reactivity was observed with Angiotensin I and Angiotensinogen. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction