Learn More
Invitrogen™ Angiotensinogen Monoclonal Antibody (1B1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515531
Description
MA5-15531 targets AGT in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15531 immunogen is purified recombinant fragment of human AGT expressed in E. Coli. MA5-15531 detects AGT which has a predicted molecular weight of approximately 53kDa.
The protein encoded by AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.
Specifications
Angiotensinogen | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P01019 | |
AGT | |
Purified recombinant fragment of human AGT expressed in E. Coli. | |
RUO | |
183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1B1 | |
Unconjugated | |
AGT | |
AGT; AI265500; alpha-1 antiproteinase, antitrypsin; Ang; Ang I; Ang II; Ang III; Ang IV; AngI; AngII; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; angiotensin II; Angiotensin III; Angiotensin IV; angiotensin ll; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (PAT); angiotensinogen (serpin peptidase inhibitor, clade A, member 8); ANHU; ANRT; Aogen; Des-Asp[1]-angiotensin II; PAT; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; serpin A8; serpin peptidase inhibitor, clade A, member 8; Serpina8 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.