Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiotensinogen Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 119921AP150UL

Description
AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in AGT gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in AGT gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.Specifications
Angiotensinogen | |
Polyclonal | |
Unconjugated | |
AGT | |
AGT, Ang I Angiotensin 2, Ang II Angiotensin 3, Ang III, Angiotensin, Angiotensin I, Angiotensin II, Angiotensin III, Angiotensinogen, ANHU, Des Asp[1] angiotensin II, Serpin A8, SERPINA8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11606, 183 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01019, P11859 | |
AGT | |
Angiotensinogen Fusion Protein Ag2632 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction