Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANGPTL1 Monoclonal Antibody (OTI9E5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA806289
Description
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.Specifications
| ANGPTL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95841 | |
| ANGPTL1 | |
| Human recombit protein fragment corresponding to amino acids 24-491of human ANGPTL1 produced in SF9 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9E5 | |
| Unconjugated | |
| ANGPTL1 | |
| ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9068 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction