Learn More
Invitrogen™ ANGPTL2 Monoclonal Antibody (239829)

Description
This antibody shows 25-100% cross-reactivity with recombinant mouse (rm) Angiopoietin-like 2 and no cross-reactivity with rhAngiopoietin-1 or rhAngiopoietin-2. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | ANGPTL2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 239829 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ANGPTL2 |
| Gene Accession No. | Q9UKU9 |
| Gene Alias | AI593246; angiopoietin like 2; angiopoietin related protein 2; angiopoietin-like 2; angiopoietin-like protein 2; Angiopoietin-related protein 2; angiopoietin-related protein-2; Angptl2; ARP2; AW260363; HARP; MGC8889; UNQ170/PRO196 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.