Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANGPTL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547139
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547139 Supplier Invitrogen™ Supplier No. PA547139
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 100% cross-reactivity with recombinant human (rh) ANGPTL2 is observed, and less than 1% cross-reactivity with rhAngiopoietin-1, rhAngiopoietin-2, recombinant mouse (rm) Angiopoietin-3, rhAngiopoietin-4, rmAngiopoietin-6, and rmAngiopoietin-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ANGPTL2
Gene Accession No. Q9R045
Gene Alias AI593246; angiopoietin like 2; angiopoietin related protein 2; angiopoietin-like 2; angiopoietin-like protein 2; Angiopoietin-related protein 2; angiopoietin-related protein-2; Angptl2; ARP2; AW260363; HARP; MGC8889; UNQ170/PRO196
Gene Symbols ANGPTL2
Host Species Goat
Immunogen E. coli-derived recombinant mouse Angiopoietin-like 2 Pro23-His493.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26360
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.