Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANGPTL2 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABC85 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC85 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC85 Supplier MilliporeSigma Supplier No. ABC85
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-ANGPTL2 antibody is validated for use in WB, IH for the detection of ANGPTL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Angiopoietin-related protein 2;Angiopoietin-like protein 2
Gene Symbols ANGPTL2;ARP2
Host Species Rabbit
Immunogen N-Trx, C-6XHis-tagged recombinant protein corresponding to human ANGPTL2.
Molecular Weight of Antigen ∽60 kDa observed
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_036230
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.